Nalgene™ Oak Ridge High-Speed PPCO Centrifuge Tubes, 50mL, With polypropylene screw caps; Capacity: 50mL - FAQs
View additional product information for Nalgene™ Oak Ridge High-Speed PPCO Centrifuge Tubes - FAQs (3119-0050, 3119-0030, 3119-0050PK, 3119-0010, 3119-0010PK, 3119-0030PK, 3119-0028, 3119-0028PK)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.