Nalgene™ PPCO Centrifuge Bottles, Capacity: 1000mL - FAQs
Nalgene™ PPCO Centrifuge Bottles, Capacity: 1000mL - FAQs
View additional product information for Nalgene™ PPCO Centrifuge Bottles - FAQs (3120-0250, 3120-1010, 3120-0250PK, 3120-0500PK, 3120-0500, 3120-9500PK, 3120-9500, 3120-1000, 3120-1000PK, 3120-1010PK)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.