Nalgene™ Opaque Amber HDPE Diagnostic Bottles with Closure: Bulk Pack
Thermo Scientific™

Nalgene™ Opaque Amber HDPE Diagnostic Bottles with Closure: Bulk Pack

Safely store and ship light-sensitive, small-volume valuable reagents with these opaque amber HDPE diagnostic bottles, for use in test kits and many small-volume applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Neck Finish
312004-90258 mL20-415 mm
312004-91254 mL13-415 mm
312004-905015 mL20-415 mm
Catalog number 312004-9025
Price (EUR)
2 156,00
Case of 2000
Capacity (Metric):
8 mL
Neck Finish:
20-415 mm
Price (EUR)
2 156,00
Case of 2000

Safely store and ship light-sensitive, small-volume valuable reagents with Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles. For use in test kits and many small-volume applications, these high density polyethylene bottles include linerless opaque amber polypropylene closures that ensures performance without wrinkles that can cause leaks or contamination of your solutions. The durable, nonpyrogenic and noncytotoxic uniform walls are resistant to splitting or puncturing.

Features:
  • Opaque amber high density polyethylene bottles with opaque amber polypropylene closures
  • Nonsterile
  • Leakproof† when used with Nalgene closures
  • Linerless caps included
  • Uniform walls that are resistant to splitting or puncturing
  • Bulk-packed for packaging applications

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton. Packed 2,000 per case.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Capacity0.25 oz., 8 mL
Capacity (English)0.25 oz.
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition.
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size20-415 mm
Closure TypeScrew Cap
Diameter (Metric)25 mm
Diameter (Metric) Inner Neck14 mm
Height (English)1.58 in. (With Closure), 1.52 in. (Without Closure)
Height (Metric)45 mm
Neck Finish20-415 mm
No. per Case2000
No. per Pack2000
Product TypeSample Container
ShapeRound
Capacity (Metric)8 mL
Closure IncludedYes
ColorOpaque Amber
GraduatedNo
MaterialHDPE
MouthNarrow
SterilityNon-sterile
Unit SizeCase of 2000