Nalgene™ Boston Round Narrow-Mouth Natural PPCO Bottles with Closure: Bulk Pack, 1000mL - FAQs
Nalgene™ Boston Round Narrow-Mouth Natural PPCO Bottles with Closure: Bulk Pack, 1000mL - FAQs
View additional product information for Nalgene™ Boston Round Narrow-Mouth Natural PPCO Bottles with Closure: Bulk Pack - FAQs (312006-0016, 312006-0008, 312006-0002, 312006-0001, 312006-0004, 312006-0032)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.