Nalgene™ Natural PPCO Diagnostic Bottles with Closure: Bulk Pack, 4mL - FAQs
Nalgene™ Natural PPCO Diagnostic Bottles with Closure: Bulk Pack, 4mL - FAQs
View additional product information for Nalgene™ Natural PPCO Diagnostic Bottles with Closure: Bulk Pack - FAQs (312006-9050, 312006-9025, 312006-9125)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.