Thermo Scientific™
Cytospin™ 4 Cytocentrifuge: Clipped Funnel Starter Kit
 Related applications:
Anatomical Pathology
For use with the Cytospin™ 4 Cytocentrifuge. Gain all the advantages of the ultimate thin-layer cell preparation system with the Thermo Scientific™ Cytospin™ 4 Cytocentrifuge. This reliable benchtop centrifuge provides economical thin-layer preparations from any liquid matrix, especially hypocellular fluids such as spinal fluid and urine.
Clipped Funnel Starter Kit includes: 50 Single White Cytofunnels, 25 Double Cytofunnels, Cytoslides, 12 Cytoclips
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
For use with the Cytospin™ 4 Cytocentrifuge. Gain all the advantages of the ultimate thin-layer cell preparation system with the Thermo Scientific™ Cytospin™ 4 Cytocentrifuge. This reliable benchtop centrifuge provides economical thin-layer preparations from any liquid matrix, especially hypocellular fluids such as spinal fluid and urine.
Clipped Funnel Starter Kit includes: 50 Single White Cytofunnels, 25 Double Cytofunnels, Cytoslides, 12 Cytoclips
Documents
Manuals & protocols