Thermo Scientific™
Shandon™ Tissue-Marking Dyes
Related applications:
Anatomical Pathology
Permanently mark the margins of excised surgical specimens for exact orientation with Thermo Scientific™ Shandon™ Tissue-Marking Dyes.
Back to top
  • Special binding pigment evenly coats the tissue without penetrating tissue surface
  • Does not interfere with diagnostic interpretation
  • Quick-drying and easy-to-use; decreases time-factor in urgent frozen-sectioning
Ordering Information:
Available individually or as a kit (includes one bottle each of black, blue, yellow, red, and green; 50 applicator sticks; storage base).