Thermo Scientific™
Shandon™ Tissue-Marking Dyes
 Related applications:
Anatomical Pathology
Permanently mark the margins of excised surgical specimens for exact orientation with Thermo Scientific™ Shandon™ Tissue-Marking Dyes.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Special binding pigment evenly coats the tissue without penetrating tissue surface
  • Does not interfere with diagnostic interpretation
  • Quick-drying and easy-to-use; decreases time-factor in urgent frozen-sectioning
Ordering Information:
Available individually or as a kit (includes one bottle each of black, blue, yellow, red, and green; 50 applicator sticks; storage base).
Documents
Manuals & protocols