Nalgene™ Square Natural PPCO Graduated Bottles with Closure: Bulk Pack, 250mL - FAQs
Nalgene™ Square Natural PPCO Graduated Bottles with Closure: Bulk Pack, 250mL - FAQs
View additional product information for Nalgene™ Square Natural PPCO Graduated Bottles with Closure: Bulk Pack - FAQs (312016-0250)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.