Nalgene™ Translucent Amber HDPE Diagnostic Bottles with Closure: Bulk Pack
Nalgene™ Translucent Amber HDPE Diagnostic Bottles with Closure: Bulk Pack
Nalgene™ Translucent Amber HDPE Diagnostic Bottles with Closure: Bulk Pack
Nalgene™ Translucent Amber HDPE Diagnostic Bottles with Closure: Bulk Pack
Thermo Scientific™

Nalgene™ Translucent Amber HDPE Diagnostic Bottles with Closure: Bulk Pack

Protect light-sensitive samples with translucent amber HDPE diagnostic bottles, intended for storage and shipping of small-volume, light-sensitive, valuable reagents.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용량(미터법)넥 마감
312084-905015 mL20-415 mm
312084-91254 mL13-415 mm
312084-90258 mL20-415 mm
카탈로그 번호 312084-9050
제품 가격(KRW)
1,660,000
Case of 2000
용량(미터법):
15 mL
넥 마감:
20-415 mm
제품 가격(KRW)
1,660,000
Case of 2000

Protect light-sensitive samples with Thermo Scientific™ Nalgene™ Translucent Amber HDPE Diagnostic Bottles with Closure, suitable for storage and shipping of small-volume, light-sensitive, valuable reagents. Translucent bottles provide light resistance with the ability to see liquid level and are ideal for test kits and many small-volume applications.

Features:
  • Nonsterile, high-density polyethylene bottles
  • Opaque, linerless polypropylene closures
  • Durable, uniform walls that resist splitting and puncturing
  • Non-cytotoxic
  • Leakproof† when used with Nalgene closures

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton; 2,000 per case.

A sample product certificate providing cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

사양
용량0.5 oz., 15 mL
용량(영국식 단위)0.5 oz.
인증/적합성Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition.
마개 라이너Unlined
설명15mL, 20-415 closure, bulk pack
직경(미터법)25 mm
직경(미터법) 내부 넥14 mm
용도(애플리케이션)Storage and Shipping of Small Volume, Light Sensitive, Valuable Reagents
높이(영국식 단위)2.28 in.
높이(미터법)58 mm
넥 마감20-415 mm
케이스당 수2000
팩당 수2000
제품 유형Sample Container
마개 디스펜싱을 통해No
용량(미터법)15 mL
Closure IncludedYes
색상Translucent Amber
물질HDPE
마우스Narrow
멸균Non-sterile
Unit SizeCase of 2000