Nalgene™ Narrow-Mouth PPCO Packaging Bottles with Closure: Bulk Pack, 500mL - FAQs
View additional product information for Nalgene™ Narrow-Mouth PPCO Packaging Bottles with Closure: Bulk Pack - FAQs (312087-0008, 312087-0002, 312087-0004, 312087-0016, 312087-0032, 312087-0001)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.