Nalgene™ Narrow-Mouth Natural HDPE Packaging Bottles with Closure: Bulk Pack
Nalgene™ Narrow-Mouth Natural HDPE Packaging Bottles with Closure: Bulk Pack
Nalgene™ Narrow-Mouth Natural HDPE Packaging Bottles with Closure: Bulk Pack
Nalgene™ Narrow-Mouth Natural HDPE Packaging Bottles with Closure: Bulk Pack
Thermo Scientific™

Nalgene™ Narrow-Mouth Natural HDPE Packaging Bottles with Closure: Bulk Pack

Store, ship and package specialty chemicals, diagnostics, biologicals, reagents, adhesives and veterinary pharmaceuticals with these narrow-mouth HDPE bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
312089-0008250 mL24-415 mm
312089-000130 mL20-415 mm
312089-000260 mL20-415 mm
312089-0004125 mL24-415 mm
312089-0016500 mL28-415 mm
312089-00321000 mL38-430 mm
Catalog number 312089-0008
Price (EUR)
229,73
Online Exclusive
277,00
Save 47,27 (17%)
Case of 250
Capacity (Metric):
250 mL
Closure Size:
24-415 mm
Price (EUR)
229,73
Online Exclusive
277,00
Save 47,27 (17%)
Case of 250

Safely store, ship and package liquid reagents using Thermo Scientific™ Nalgene™ Narrow-Mouth Natural HDPE Packaging Bottles with Closure, an ideal choice for specialty chemicals, diagnostics, biologicals, reagents, adhesives and veterinary pharmaceuticals. The injection blow-molded high-density polyethylene bottles feature a tough plastic construction with uniform wall thickness and strength. Polypropylene closures are included for leakproof† sealing.

Features:
  • Made from high-purity, food-grade resins
  • Recommended for storage and shipment of liquid reagents

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton.

.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Capacity8 oz., 250 mL
Closure ColorNatural, translucent
Closure MaterialPolypropylene
Closure Size24-415 mm
Closure TypeScrew (Bulk Packed, Unassembled)
DescriptionNarrow-Mouth Packaging Bottles
Diameter (Metric)61 mm
Diameter (Metric) Inner Neck18 mm
Height (English)2.4 in.
Height (Metric)133 mm
Neck Finish24 mm
No. per Case250
No. per Pack250
Through the Closure DispensingNo
Width (English)2.4 in.
Capacity (English)8 oz.
Capacity (Metric)250 mL
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
ColorTranslucent
MaterialHDPE
MouthNarrow
Product TypePacker Bottle
ShapeRound
Unit SizeCase of 250