Nalgene™ Narrow-Mouth HDPE IP2 Bottles with Closure: Bulk Pack
Thermo Scientific™

Nalgene™ Narrow-Mouth HDPE IP2 Bottles with Closure: Bulk Pack

Ship and store liquid reagents using these high density, durable polyethylene bottles with closure. Recommended for customers who design, assemble and certify their own combination packaging.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeUnit Size
312099-00321000 mL38-430 mmCase of 50
312099-000130 mL20-415 mmCase of 1000
312099-000260 mL20-415 mmCase of 1000
312099-0004125 mL24-415 mmCase of 500
312099-0008250 mL24-415 mmCase of 250
312099-0016500 mL28-415 mmCase of 125
312099-0016TS
also known as 312099-0016
500 mL28-415 mmCase of 125
Catalog number 312099-0032
Price (USD)
-
Capacity (Metric):
1000 mL
Closure Size:
38-430 mm
Unit Size:
Case of 50

Ship and store liquid reagents using durable Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE IP2 Bottles with Closure. These high density polyethylene bottles are a safe, economical solution for customers who design, assemble and certify their own combination packaging.

Features:
  • HDPE bottle with PP closure, bulk pack
  • Narrow mouth
  • Complies with IATA and ICAO regulations

Ordering Information:

Bottles and closures are bulk packed in separate polybags within the same carton

Recommended for:

Storage and shipment of liquid reagents

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Capacity32 oz., 1000 mL
Capacity (English)32 oz.
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27 (c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9.
Closure MaterialPolypropylene
Closure Size38-430 mm
DescriptionIP2 Narrow-Mouth Bottles
Diameter (Metric)91 mm
Diameter (Metric) Inner27 mm
Diameter (Metric) Inner Neck27 mm
For Use With (Application)Storage and Shipment of Liquid Reagents
Height (English)8.5 in.
Height (Metric)216 mm
Height (Metric) With Closure216 mm
No. per Case50
No. per Pack50
Product TypeIP2 Bottle
ShapeRound
Capacity (Metric)1000 mL
Closure IncludedYes
ColorTranslucent
MaterialHDPE
MouthNarrow
Unit SizeCase of 50