Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.
Search
View additional product information for Nalgene™ Wide-Mouth PPCO Bottles with Closure: Bulk Pack, Autoclavable - FAQs (312105-0002)
1 product FAQs found