Nalgene™ Wide-Mouth PPCO Bottles with Closure: Bulk Pack, Autoclavable, 1000mL - FAQs
Nalgene™ Wide-Mouth PPCO Bottles with Closure: Bulk Pack, Autoclavable, 1000mL - FAQs
View additional product information for Nalgene™ Wide-Mouth PPCO Bottles with Closure: Bulk Pack, Autoclavable - FAQs (312105-0008, 312105-0016, 312105-0032, 312105-0004, 312105-0001)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.