Nalgene™ Square Wide-Mouth PPCO Bottles with Closure: Bulk Pack, Autoclavable, 175mL - FAQs
Nalgene™ Square Wide-Mouth PPCO Bottles with Closure: Bulk Pack, Autoclavable, 175mL - FAQs
View additional product information for Nalgene™ Square Wide-Mouth PPCO Bottles with Closure: Bulk Pack, Autoclavable - FAQs (312110-0006)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.