Heat-Shrink Bands for Nalgene™ PETG Media Bottles
Actual product may vary
Thermo Scientific™
Heat-Shrink Bands for Nalgene™ PETG Media Bottles
Ensure the integrity of bottle contents with these tamper-resistant seals. Heat-shrink bands are gamma stable and include a perforated tear strip feature for easy removal.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNeck Finish
312160-053053B
312160-020020-415
312160-024024-415
312160-038438-430
Catalog number 312160-0530
Price (KRW)
-
Neck Finish:
53B
Ensure the integrity of bottle contents with Thermo Scientific™ Heat-Shrink Bands for Thermo Scientific™ Nalgene™ PETG Media Bottles. Simply apply the recommended application torque for Thermo Scientific Nalgene closures and slide the heat-shrink band over the closure and bottle neck. Heat the band to shrink and secure contents with a tamper-resistant seal.
  • Gamma stable bands include a perforated ”tear strip” feature for easy removal.
  • Bands available are in four sizes to mate with Nalgene PETG Square Media Bottles.
  • Bands are packed 1,000 per case (two zipper bags of 500 each).
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Warranty: 90 days
Specifications
For Use With (Equipment)2000 mL Square Media Bottle
Neck Finish53B
No. per Case1000
No. per Pack1000
Product TypeHeat-shrink Band
Torque38-53 in.-lb
Torque Wrench Fittings2195-1153
MaterialPETG
Unit SizeCase of 1000