Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure: Bulk Pack
Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure: Bulk Pack
Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure: Bulk Pack
Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure: Bulk Pack
Thermo Scientific™

Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure: Bulk Pack

Ship and store diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals in these light-weight bottles which are leakproof when used with Nalgene closures.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용량(미터법)마개 크기
312187-0004125 mL38-415 mm
312187-000130 mL28-415 mm
312187-000260 mL28-415 mm
312187-0008250 mL43-415 mm
312187-0016500 mL53-415 mm
312187-00321000 mL63-415 mm
카탈로그 번호 312187-0004
제품 가격(KRW)
603,000
Case of 500
용량(미터법):
125 mL
마개 크기:
38-415 mm
제품 가격(KRW)
603,000
Case of 500

Ship and store diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals with Thermo Scientific™ Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure. These durable, autoclavable bottles are designed for shipping and storage.

Features:
  • Wide mouth for easy filling and dispensing of powders
  • Injection blow molded for uniform wall thickness and strength
  • Made from high-purity, food-grade resins; no plasticizers, fillers or extenders
  • Polypropylene closures included
  • Leakproof† when used with Nalgene closures

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

사양
오토클레이브 가능Autoclavable
용량4 oz., 125 mL
용량(영국식 단위)4 oz.
인증/적합성Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
마개 색상Natural, translucent
마개 재질Polypropylene
마개 크기38-415 mm
마개 유형Screw (Bulk Packed, Unassembled)
설명Wide-Mouth Packaging Bottles
직경(미터법)50 mm
직경(미터법) 내부 넥28 mm
높이(영국식 단위)3.92 in. (With Closure); 3.78 in. (Without Closure)
높이(미터법)99 mm
넥 마감38 mm
케이스당 수500
팩당 수500
제품 유형Packer Bottle
형상Round
폭(영국식 단위)1.99 in.
용량(미터법)125 mL
Closure IncludedYes
색상Translucent
GraduatedNo
물질PPCO
마우스Wide
Unit SizeCase of 500

자주 묻는 질문(FAQ)

Are the Nalgene PPCO media bottles/tubes able to be irradiated?

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.