Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure: Bulk Pack, 500mL - FAQs
View additional product information for Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure: Bulk Pack - FAQs (312187-0008, 312187-0004, 312187-0016, 312187-0032, 312187-0002, 312187-0001)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.