Product Image
Gibco™

Waymouth's Medium

Waymouth's Medium is a chemically defined medium developed by Charity Waymouth and originally formulated for studies on the nutrition, metabolism,Read more
Have Questions?
Catalog NumberQuantity
3122007210 x 500 mL
Catalog number 31220072
Price (MXN)
-
Quantity:
10 x 500 mL
Customize this product

Waymouth's Medium is a chemically defined medium developed by Charity Waymouth and originally formulated for studies on the nutrition, metabolism, and growth of mouse Strain L sublines, NCTC clone 929 (L929 cells) in a serum-free environment. Gibco™ Waymouth's Medium is also used for the culture of whole organs and a broad range of cell types, such as carcinoma cell lines.

This Waymouth's Medium is modified as follows:
WithWithout
• L-glutamine• HEPES
• Phenol Red 


The complete formulation is available.

Waymouth's Medium contains no proteins, lipids, or growth factors. Some cell types may required the supplementation with Fetal Bovine Serum (FBS). Waymouth's Medium uses a sodium bicarbonate buffer system (2.24 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Specifications
Cell TypeMammalian Cells
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeWaymouth's Medium
Quantity10 x 500 mL
Shelf Life12 Months
ClassificationAnimal Origin-free
Culture TypeMammalian Cell Culture
FormLiquid
SterilitySterile-filtered
With AdditivesGlutamine, Phenol Red
Without AdditivesNo HEPES
Unit SizeEach
Contents & Storage
Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture