(R)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 99%
(R)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 99%
(R)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 99%
Thermo Scientific Chemicals

(R)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 99%

CAS: 20445-31-2 | C10H9F3O3 | 234.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number 312222500
Price (USD)
57.65
Special offer
Online exclusive
Ends: 28-Dec-2026
67.50
Save 9.85
Each
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
57.65
Special offer
Online exclusive
Ends: 28-Dec-2026
67.50
Save 9.85
Each
Chemical Identifiers
CAS20445-31-2
IUPAC Name3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid
Molecular FormulaC10H9F3O3
InChI KeyJJYKJUXBWFATTE-UHFFFAOYNA-N
SMILESCOC(C(O)=O)(C1=CC=CC=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Titration with NaOH98.5 to 101.5 %
Appearance (Form)Viscous liquid or low melting solid
Infrared spectrumConforms
Specific optical rotation+71° to +74° (20°C, 589 nm) (c=2, CH3OH)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagent
RUO – Research Use Only