2-(Trifluoromethyl)benzylamine, 98%
2-(Trifluoromethyl)benzylamine, 98%
2-(Trifluoromethyl)benzylamine, 98%
Thermo Scientific Chemicals

2-(Trifluoromethyl)benzylamine, 98%

CAS: 3048-01-9 | C8H8F3N | 175.16 g/mol
数量:
1 g
5 g
10 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 312950100
価格(JPY)
-
数量:
10 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3048-01-9
IUPAC Name1-[2-(trifluoromethyl)phenyl]methanamine
Molecular FormulaC8H8F3N
InChI KeyZSKQIFWUTUZAGF-UHFFFAOYSA-N
SMILESNCC1=CC=CC=C1C(F)(F)F
さらに表示
Refractive index1.4700 to 1.4720 (20°C, 589 nm)
Titration with HClO4>=97.5 %
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only