3-(Trifluoromethyl)benzylamine, 98%
3-(Trifluoromethyl)benzylamine, 98%
3-(Trifluoromethyl)benzylamine, 98%
Thermo Scientific Chemicals

3-(Trifluoromethyl)benzylamine, 98%

CAS: 2740-83-2 | C8H8F3N | 175.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3129600505 gGlass bottle
Catalog number 312960050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2740-83-2
IUPAC Name1-[3-(trifluoromethyl)phenyl]methanamine
Molecular FormulaC8H8F3N
InChI KeyYKNZTUQUXUXTLE-UHFFFAOYSA-N
SMILESNCC1=CC=CC(=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.4620 to 1.4640 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only