Tris(pentafluorophenyl)phosphine, 97%
Tris(pentafluorophenyl)phosphine, 97%
Tris(pentafluorophenyl)phosphine, 97%
Thermo Scientific Chemicals

Tris(pentafluorophenyl)phosphine, 97%

CAS: 1259-35-4 | C18F15P | 532.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 313160050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1259-35-4
IUPAC Nametris(2,3,4,5,6-pentafluorophenyl)phosphane
Molecular FormulaC18F15P
InChI KeyFQLSDFNKTNBQLC-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(P(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to beige to light brown
Melting point108°C to 119°C
Appearance (Form)Powder or crystals
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only