Search
View additional product information for DMEM/F-12, HEPES - FAQs (31330038)
2 product FAQs found
The color change of the lid has no effect on the sterility or functionality of the product. There has been no change to the end product DMEM/F-12, HEPES medium (Cat. No. 31330038, 11330032, 31330095, 11330057, 11330107); the medium still has the same composition.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
We do not offer DMEM/F12 medium without folic acid as a standard catalog product. However, our Customs group can make it for you as a custom media. If you are interested, please complete this Custom order inquiry form PDF (https://www.thermofisher.com/content/dam/LifeTech/global/applied-sciences/pdfs/Bioproduction/Gibco-Custom-Media-Buffers-Request-Form.pdf) and email it to custommedia@thermofisher.com. Our Customs group will review your requirements and get back to you within a couple of days with pricing, ordering information, and lead time. After that, you have the choice of proceeding with an official order or declining.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.