Pierce™ Alkaline Phosphatase

Catalog number: 31392B

Thermo Scientific™  Related applications: Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This purified calf intestinal alkaline phosphatase (CIP) is supplied in Tris buffer and 50% glycerol for use in protein research methods. The main applications for alkaline phosphatase in molecular biology and protein research are to remove 5'-phosphate groups from DNA or as a reporter system for immunoassays such as ELISA. For these latter methods, the enzyme is usually conjugated to specific primary or secondary antibodies and its activity is detected with a color-forming (or light-generating) substrate.

Features of Alkaline Phosphatase:

Purified form—ready to dilute and conjugate without prior dialysis
Concentrated—approximately 20 mg/mL (lot-specific value reported)
High specific activity—typically greater than 1600 units/mg (lot-specific value reported)
Tris buffered solution—supplied in 5 mM Tris, 5 mM magnesium chloride and 0.1 mM zinc chloride,
pH ~7.0 in 50% glycerol

One unit equals the amount of protein required to hydrolyze one micromole of p-nitrophenyl phosphate (PNPP) per minute at 25°C in a glycine buffer, pH 9.6.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: AP (Alkaline Phosphatase)
Product Line: Pierce™
Product Size: Custom

Contents & storage

Store at 2–8°C.

Documents

Manuals & protocols