Nalgene™ Polycarbonate Centrifuge Bottles with Sealing Closure
Nalgene™ Polycarbonate Centrifuge Bottles with Sealing Closure
Nalgene™ Polycarbonate Centrifuge Bottles with Sealing Closure
Nalgene™ Polycarbonate Centrifuge Bottles with Sealing Closure
Thermo Scientific™

Nalgene™ Polycarbonate Centrifuge Bottles with Sealing Closure

Process large-volume cell harvesting, pelleting and protein purification with these polycarbonate centrifuge bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNominal CapacityUnit Size
3140-0500PK
also known as 3140-0500
450 mLPack of 4
3140-0250250 mLCase of 36
3140-0250PK
also known as 3140-0250
250 mLPack of 4
3140-0500450 mLCase of 24
Catalog number 3140-0500PK
also known as 3140-0500
Price (EUR)
158,00
Pack of 4
Nominal Capacity:
450 mL
Unit Size:
Pack of 4
Price (EUR)
158,00
Pack of 4

Ideal for large-volume processes such as cell harvesting, pelleting, and protein purification, Thermo Scientific™ Nalgene™ PC Centrifuge Bottles are molded of crystal-clear polycarbonate, which allows easy sample visualization and results in excellent mechanical strength. These centrifuge bottles are the perfect fit with Thermo Scientific™ Fiberlite™ carbon fiber rotors.

  • Molded of polycarbonate for easy visualization of samples, these bottles are ideal for low- to moderate-speed centrifugation of biological and chemical samples
  • Polypropylene sealing cap closure with silicone gasket adds leakproof† assurance at maximum speeds or when spinning hazardous materials
  • Fill bottles to 80% of total capacity
  • Autoclavable; Repeated autoclaving will shorten the life of PC centrifuge bottles. 

Ordering Information:
When spinning 250mL bottle in a Thermo Scientific™ Sorvall™ GSA rotor, use of the Nalgene centrifuge bottle adapter (DS3125-0250) is required.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
AutoclavableAutoclavable
Capacity (English)16.9 oz.
Closure MaterialPolypropylene, red rubber
Closure TypeSealing Cap w/Silicone Gasket
Diameter (Metric)69.5 mm
Height (English)6.29 in.
Height (Metric)159.5 mm
Length (English)2.735 in.
Nominal Capacity450 mL
No. per Case24
No. per Pack4
Product TypeCentrifuge Bottle
ShapeRound
Through the Closure DispensingNo
Width (English)2.735 in.
Capacity (Metric)450 mL
Closure IncludedYes
ColorClear
GraduatedNo
MaterialPolycarbonate
Max. Force13,700 x g
Unit SizePack of 4