Nalgene™ PPCO Centrifuge Bottles with Sealing Closure
Nalgene™ PPCO Centrifuge Bottles with Sealing Closure
Nalgene™ PPCO Centrifuge Bottles with Sealing Closure
Nalgene™ PPCO Centrifuge Bottles with Sealing Closure
Thermo Scientific™

Nalgene™ PPCO Centrifuge Bottles with Sealing Closure

Centrifugalize low- to moderate-speed, large-volume, biological and chemical samples with these PPCO centrifuge bottles, which come with a sealing closure.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoNominal Capacity
3141-0500450 mL
3141-0250250 mL
Número do catálogo 3141-0500
Preço (BRL)
-
Nominal Capacity:
450 mL

Centrifugalize low- to moderate-speed, large-volume, biological and chemical samples with Thermo Scientific™ Nalgene™ PPCO Centrifuge Bottles, which include a sealing closure for added leakproof† assurance at maximum speeds when spinning hazardous materials. Molded of rigid and robust polypropylene copolymer, these translucent bottles are designed for pelleting applications, such as cell culture harvesting and protein biochemistry, including ammonium sulfate precipitations. Additionally, these PPCO bottles are autoclavable and offer excellent mechanical strength.

Features:
  • For low- to moderate-speed centrifugation of biological and chemical samples
  • Bottle includes polypropylene sealing cap assembly with silicone gasket for added leakproof† assurance at maximum speeds or when spinning hazardous materials
  • For proper performance, bottles must be filled to 80% of total capacity
  • Autoclavable

Requires:

When spinning 250mL bottle in a Sorvall GSA rotor, use of the Nalgene centrifuge bottle adapter (DS3125-0250) is required.

Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) centrifuge tubes are designed to last many spins under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. These centrifuge tubes are recyclable in many communities. At end of product life, please wash and recycle your Nalgene PP and PPCO centrifuge tubes if services are available in your community.

Especificações
AutoclavableAutoclavable
Capacity (English)16.9 oz.
Closure MaterialPolypropylene, red rubber
Closure TypeSealing Cap Assembly with Silicone Gasket
Diameter (Metric)69.5 mm
Height (English)6.27 in.
Height (Metric)159.3 mm
Length (English)2.735 in.
Length (Metric)160 mm
Nominal Capacity450 mL
No. per Pack4
Product TypeCentrifuge Bottle
ShapeRound
Through the Closure DispensingNo
Width (English)2.735 in.
Capacity (Metric)450 mL
Closure IncludedYes
ColorTranslucent
GraduatedNo
MaterialPPCO
Max. Force13,700 x g
Unit SizeCase of 24

Frequently asked questions (FAQs)

Are the Nalgene PPCO media bottles/tubes able to be irradiated?

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.