Nalgene™ PPCO Centrifuge Bottles with Sealing Closure, 450mL - FAQs
View additional product information for Nalgene™ PPCO Centrifuge Bottles with Sealing Closure - FAQs (3141-0250, 3141-0250PK, 3141-0500PK, 3141-0500)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.