1,4-Diisocyanatobutane, 97%
1,4-Diisocyanatobutane, 97%
1,4-Diisocyanatobutane, 97%
Thermo Scientific Chemicals

1,4-Diisocyanatobutane, 97%

CAS: 4538-37-8 | C6H8N2O2 | 140.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 314380050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4538-37-8
IUPAC Name1,4-diisocyanatobutane
Molecular FormulaC6H8N2O2
InChI KeyOVBFMUAFNIIQAL-UHFFFAOYSA-N
SMILESO=C=NCCCCN=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4510 to 1.4530 (20°C, 589 nm)
Appearance (Color)Clear light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only