p-Tolyl chloroformate, 97%
p-Tolyl chloroformate, 97%
p-Tolyl chloroformate, 97%
Thermo Scientific Chemicals

p-Tolyl chloroformate, 97%

CAS: 937-62-2 | C8H7ClO2 | 170.6 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3145600505 gGlass bottle
Catalog number 314560050
Price (USD)
46.65
Special offer
Online exclusive
Ends: 28-Dec-2026
54.60
Save 7.95
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
46.65
Special offer
Online exclusive
Ends: 28-Dec-2026
54.60
Save 7.95
Each
Chemical Identifiers
CAS937-62-2
IUPAC Name4-methylphenyl carbonochloridate
Molecular FormulaC8H7ClO2
InChI KeyXOFZPIYYMJUNRG-UHFFFAOYSA-N
SMILESCC1=CC=C(OC(Cl)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5080 to 1.5100 (20°C, 589 nm)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only