EZ-Link™ Maleimide Activated Horseradish Peroxidase

Catalog number: 31485B

Thermo Scientific™  Related applications: Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce Maleimide Activated Horseradish Peroxidase is for preparation of HRP conjugates with proteins, peptides or other ligands that contain sulfhydryl groups, such as reduced cysteines. This product is for bulk orders. Thermo Scientific Pierce Maleimide Activated Horseradish Peroxidase and can effectively modify an equivalent amount of immunoglobulin (1 mg/mg IgG).

Features of Maleimide Activated Horseradish Peroxidase:

Activated HRP – horseradish peroxidase (HRP) modified with maleimide groups for conjugation to sulfhydryl molecules
Sulfhydryl-reactivemaleimide groups conjugate to reduced thiols (-SH), as in the side-chain of cysteine residues
High activity HRP – enzyme activity is greater than 240 units/mg; lyophilized, activated enzyme is stable for at least 12 months at 4°C
Complete kit – includes the activated HRP as well as two types of reagents for sulfhydryl-ready antibodies (IgG) or proteins for conjugation

This product consists of horseradish peroxidase (HRP) that has been modified with Sulfo-SMCC (Part No. 22322) to attach several maleimide groups per HRP molecule while retaining the peroxidase activity. The activated HRP will covalently attach to proteins or other molecule containing sulfhydryl groups (e.g., cysteines). HRP-conjugates of antibodies, proteins, peptides and other thiol-containing reporter probes are easily made using this method. The complete kit includes the activated HRP as well as two types of reagents for preparing sulfhydryl-ready antibodies (IgG) or proteins for conjugation.

The complete kit for Maleimide Activated Horseradish Peroxidase contains reagents for exposing or added the necessary sulfhydryl groups on antibodies (IgG) or practically any other protein. These general strategies are described briefly in the applications section of our review of Maleimide Reaction Chemistry. Of course, any protein that contains cysteines has sulfhydryl groups (-SH), but they must be reduced (not in the form of disulfide bonds) to be conjugated. Antibodies also contain disulfide bonds that can be targeted as antibody labeling sites; the hinge-region disulfide bonds in IgG can be selectively cleaved with the mild reducing agent 2-Mercaptoethylamine (Part No. 20408), which is included in the complete kit. Alternatively, sulfhydryl groups can be added to proteins (or any amine-containing molecule) using SATA reagent (Part No. 26102), which also is included in the kit.

Related Products
EZ-Link™ Maleimide Activated Horseradish Peroxidase Kit
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity: Thiol
Label or Dye: HRP (Horseradish Peroxidase)
Product Line: EZ-Link™
Product Size: Custom
Solubility: Water

Contents & storage

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.

Documents

Manuals & protocols