Pierce™ Horseradish Peroxidase
Thermo Scientific™

Pierce™ Horseradish Peroxidase

Thermo Scientific Pierce Horseradish Peroxidase (HRP) is purified horseradish peroxidase enzyme for use in activity assays and conjugation to antibodies자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
3149010 mg
31491100 mg
카탈로그 번호 31490
제품 가격(KRW)
139,000
온라인 행사
Ends: 30-Sep-2026
163,000
할인액 24,000 (15%)
Each
수량:
10 mg
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
139,000
온라인 행사
Ends: 30-Sep-2026
163,000
할인액 24,000 (15%)
Each

Thermo Scientific Pierce Horseradish Peroxidase (HRP) is purified horseradish peroxidase enzyme for use in activity assays and conjugation to antibodies for ELISA, Western blot and immunohistochemistry applications.

Features of Thermo Scientific Pierce Horseradish Peroxidase:

  • Purified form—lyophilized salt-free powder; ready to dissolve and use
  • High specific activity—typically greater than 250 units/mg (lot-specific value reported)
  • Compared to AP—HRP is smaller (40kDa) than alkaline phosphatase (AP; 140kDa) and has higher specific enzyme activity than both AP and beta-galactosidase (b-Gal)
  • Many options—numerous substrate solutions and assay techniques are available for HRP

This purified horseradish peroxidase (HRP) is supplied lyophilized as a salt-free powder for reconstitution and use in protein research methods. The main application for HRP in molecular biology and protein research is as a reporter system for immunoassays and other probe-based assay techniques such as ELISA, Western blotting, EMSA and Southern blotting. The enzyme is usually conjugated to specific secondary antibodies or streptavidin, and its activity is detected with a color-forming (or light-generating) substrate.

One unit catalyses the production of 1 mg of purpurogallin from pyrogallol in 20 seconds at 20°C and pH 6.0.

For Research Use Only. Not for use in diagnostic procedures.
사양
라벨 또는 염료HRP (Horseradish Peroxidase)
수량10 mg
용도(애플리케이션)Western Blotting, ELISA, Immunohistochemistry
형태Powder
제품라인Pierce
Unit SizeEach
구성 및 보관
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C.