Erbium(III) nitrate pentahydrate, 99.9%, (trace metal basis)
Erbium(III) nitrate pentahydrate, 99.9%, (trace metal basis)
Erbium(III) nitrate pentahydrate, 99.9%, (trace metal basis)
Thermo Scientific Chemicals

Erbium(III) nitrate pentahydrate, 99.9%, (trace metal basis)

CAS: 10031-51-3 | ErH10N3O14 | 443.35 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
產品號碼 315141000
價格 (TWD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
化學識別
CAS10031-51-3
IUPAC Nameerbium(3+) pentahydrate trinitrate
Molecular FormulaErH10N3O14
InChI KeyLWHHUEHWVBVASY-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
檢視更多
規格Specification Sheet規格表
Appearance (Color)Pink
Appearance (Form)Crystalline powder or crystals
Assay33 to 39 % (Er)
Total conc. of imp.as REO=<0.1 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only