Bismuth(III) nitrate pentahydrate, 99.999%, (trace metal basis)
Bismuth(III) nitrate pentahydrate, 99.999%, (trace metal basis)
Bismuth(III) nitrate pentahydrate, 99.999%, (trace metal basis)
Thermo Scientific Chemicals

Bismuth(III) nitrate pentahydrate, 99.999%, (trace metal basis)

CAS: 10035-06-0 | BiH10N3O14 | 485.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 315150250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10035-06-0
IUPAC Namebismuth(3+) pentahydrate trinitrate
Molecular FormulaBiH10N3O14
InChI KeyFBXVOTBTGXARNA-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total trace metal impurities=<0.001 %
Appearance (Color)White
Appearance (Form)Crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only