Strontium nitrate, 99+%, ACS reagent
Strontium nitrate, 99+%, ACS reagent
Strontium nitrate, 99+%, ACS reagent
Thermo Scientific Chemicals

Strontium nitrate, 99+%, ACS reagent

CAS: 10042-76-9 | N2O6Sr | 211.63 g/mol
数量:
100 g
500 g
2.5 kg
パッケージ:
Plastic bottle
製品番号(カタログ番号) 315175000
価格(JPY)
-
数量:
500 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10042-76-9
IUPAC Namestrontium(2+) dinitrate
Molecular FormulaN2O6Sr
InChI KeyDHEQXMRUPNDRPG-UHFFFAOYSA-N
SMILES[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
さらに表示
Appearance (Color)White
Loss on drying=<0.1 % (105°C)
Heavy metals (ICP-OES)=<5 ppm
Chloride (Cl)=<20 ppm
Sodium (Na)=<1000 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only