Lanthanum chloride, 99.9%
Lanthanum chloride, 99.9%
Lanthanum chloride, 99.9%
Thermo Scientific Chemicals

Lanthanum chloride, 99.9%

CAS: 10099-58-8 | Cl3La | 245.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 315230100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10099-58-8
IUPAC Namelanthanum(3+) trichloride
Molecular FormulaCl3La
InChI KeyICAKDTKJOYSXGC-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[La+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Total conc. of imp.as REO=<0.1 %
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only