Product Image
Gibco™

Ham's F-10 Nutrient Mix

Ham's F-10 Nutrient Mixture (F-10) was designed for serum-free growth of Chinese Hamster Ovary (CHO) cells. F-10 has since beenRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
31550049
also known as 31550-049
20 x 100 mL
31550015
also known as 31550-015
100 mL
31550023
also known as 31550-023
500 mL
31550031
also known as 31550-031
10 x 500 mL
Catalog number 31550049
also known as 31550-049
Price (INR)
-
Quantity:
20 x 100 mL
Customize this product

Ham's F-10 Nutrient Mixture (F-10) was designed for serum-free growth of Chinese Hamster Ovary (CHO) cells. F-10 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including COS-7, primary rat astrocytes, and rat prostate epithelial cells.


This F-10 is modified as follows:
WithWithout
• L-Glutamine• HEPES
• Phenol Red 


The complete formulation is available.

Compared to other basal media, F-10 contains a wider variety of components, including zinc, hypoxanthine, and thymidine. Serum-free growth of CHO cells in F-10 has led to a variety of improved formulations, such as Ham's F-12 Nutrient Mixture (F-12).

Gibco™ F-10 contains no proteins or growth factors. Therefore, F-10 often requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). F-10 uses a sodium bicarbonate buffer system (1.2 g/L), and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Specifications
Cell TypeMammalian Cells
Concentration1 X
Culture EnvironmentCO2
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality275 - 315 mOsm/kg
Product LineGibco
Product TypeHam's F-10 Nutrient Mix
Quantity20 x 100 mL
Shelf Life12 Months
ClassificationAnimal Origin-free
Culture TypeMammalian Cell Culture
FormLiquid
SterilitySterile-filtered
With AdditivesGlutamine, Sodium Bicarbonate, Sodium Pyruvate
Without AdditivesNo HEPES
pH7 to 7.4
Unit SizeEach
Contents & Storage
Storage conditions: 2–8°C. Protect from light.
Shipping conditions: Ambient.
Shelf life: 12 months from date of manufacture.