Thermo Scientific Chemicals

Tellurium, 99.8%, (trace metal basis), powder, -200 mesh, Thermo Scientific Chemicals

Catalog number: 315995000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Tellurium, 99.8%, (trace metal basis), powder, -200 mesh, Thermo Scientific Chemicals

Catalog number: 315995000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 315995000
Price (USD)
Price: 510.00
Your price:
Quantity
-

Chemical Identifiers

CAS
13494-80-9
IUPAC Name
tellurium
Molecular Formula
Te
InChI Key
PORWMNRCUJJQNO-UHFFFAOYSA-N
SMILES
[Te]
Appearance (Color)
Grey to black
Total trace metal impurities
=<0.2 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text