Thermo Scientific™
Concentric Ring Cover for Precision™ General-Purpose Water Baths
Convert the Model 184 or 284 bath for use as a steaming bath with Thermo Scientific™ Precision Concentric Ring Cover.
Back to top
  • Stainless steel
  • Five openings (3 small, 2 large) accommodate glassware ranging in diameter from 2 to 10cm (0.8 to 4in.) in 2.5cm (1in.) increments
  • Openings enlarge as rings are removed