Perrhenic(VII)acid, 99.99%, (trace metal basis), 76.5% solution in water
Perrhenic(VII)acid, 99.99%, (trace metal basis), 76.5% solution in water
Perrhenic(VII)acid, 99.99%, (trace metal basis), 76.5% solution in water
Thermo Scientific Chemicals

Perrhenic(VII)acid, 99.99%, (trace metal basis), 76.5% solution in water

CAS: 13768-11-1 | HO4Re | 251.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 316180250
Price (EUR)
360,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
360,00
Each
Chemical Identifiers
CAS13768-11-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Total trace metal impurities=<100 ppm
Appearance (Color)Clear colorless to light yellow
Strength70 to 77 % (HReO4)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Reduction reagent
RUO – Research Use Only