Thermo Scientific™
Clips and Fasteners for Precision™ Water Baths
Clips and fasteners for use with Thermo Scientific™ Precision™ water baths.
Back to top
Flask Clips
  • Secure various size flasks to bath platform
  • Stainless steel
  • Each requires one fastener (Cat. No. 3166189 - order separately)
Test Tube Clip
  • Stainless steel
  • Hold 13mm to 25mm tubes
  • Each requires one fastener (Cat. No. 3166189 - order separately)
  • Warranty varies by country, please contact us for details