Search
| 產品號碼 | For Use With (Equipment) |
|---|---|
| 3166211 | Steaming Water Baths |
| 3166223 | Circulating, Shaking Water Baths |
Thermo Scientific™ Precision Water Level Regulator Kits are for use with Precision water baths.
Includes:6ft. (1.8m) 0.25in. Tygon tubing, nylon fittings, clamps and adjustable mounting bracket