Product Image
Thermo Scientific™

Glass Thermometers for Precision™ Water Baths

Use these glass thermometers with precision general-purpose and circulating water baths.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Length (Metric)
3166220All Precision baths except 180/280305 mm
3166236Models 180/280 Precision Water Bath165 mm
Catalog number 3166220
Price (USD)
-
For Use With (Equipment):
All Precision baths except 180/280
Length (Metric):
305 mm

Thermo Scientific™ Glass Thermometers are for use with Thermo Scientific™ Precision General-Purpose and Circulating Water Baths.

  • Nonhazardous spirit-filled glass thermometers read in 1.0°C increments
  • For monitoring purposes only
  • Warranty varies by country, please contact us for details
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Equipment)All Precision baths except 180/280
Length (English)12 in.
Length (Metric)305 mm
Temperature Range0°C to 100°C
TypeThermometer
Unit SizeEach