Gadolinium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Gadolinium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Gadolinium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Thermo Scientific Chemicals

Gadolinium(III) nitrate hexahydrate, 99.9%, (trace metal basis)

CAS: 19598-90-4 | GdN3O9·6H2O | 451.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 316761000
Price (USD)
306.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
306.00
Each
Chemical Identifiers
CAS19598-90-4
IUPAC Namegadolinium(3+) hexahydrate trinitrate
Molecular FormulaGdH12N3O15
InChI KeyXWFVFZQEDMDSET-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Gd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Gadolinium (Gd)34 to 36 %
Total conc. of imp.as REO=<0.1 %
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only