Cadmium acetate dihydrate, 98%, for analysis
Cadmium acetate dihydrate, 98%, for analysis
Cadmium acetate dihydrate, 98%, for analysis
Thermo Scientific Chemicals

Cadmium acetate dihydrate, 98%, for analysis

CAS: 5743-04-4 | C4H6CdO4·2H2O | 266.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 317130025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS5743-04-4
IUPAC Namecadmium(2+) diacetate dihydrate
Molecular FormulaC4H10CdO6
InChI KeyAUIZLSZEDUYGDE-UHFFFAOYSA-L
SMILESO.O.[Cd++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Nitrate (NO3)=<30 ppm
Iron (Fe)=<10 ppm
Appearance (Color)White
Titration Complexometric>=97.5 %
Subst not pptd by H2S=<0.3 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only