Silver, 99.9%, (trace metal basis), wire, 2.0 mm diam., 1ea = 16.5gr
Silver, 99.9%, (trace metal basis), wire, 2.0 mm diam., 1ea = 16.5gr
Silver, 99.9%, (trace metal basis), wire, 2.0 mm diam., 1ea = 16.5gr
Thermo Scientific Chemicals

Silver, 99.9%, (trace metal basis), wire, 2.0 mm diam., 1ea = 16.5gr

CAS: 7440-22-4 | Ag | 107.87 g/mol
製品番号(カタログ番号)数量
3177200101 Ea.
製品番号(カタログ番号) 317720010
価格(JPY)
-
数量:
1 Ea.
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
さらに表示
Appearance (Color)Silver white
Total trace metal impurities=<0.1 %
Appearance (Form)Wire
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only