RPMI 1640 Medium, powder
Gibco™

RPMI 1640 Medium, powder

RPMI 1640 Medium was originally developed to culture human leukemic cells in suspension and as a monolayer. Roswell Park Memorial자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
3180008910 L
3180002210 x 1 L
카탈로그 번호 31800089
제품 가격(KRW)
63,000
온라인 행사
Ends: 30-Sep-2026
70,000
할인액 7,000 (10%)
Each
수량:
10 L
Customize this product
제품 가격(KRW)
63,000
온라인 행사
Ends: 30-Sep-2026
70,000
할인액 7,000 (10%)
Each
RPMI 1640 Medium was originally developed to culture human leukemic cells in suspension and as a monolayer. Roswell Park Memorial Institute (RPMI) 1640 Medium has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of RPMI 1640 Medium modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This RPMI is modified as follows:
WithWithout
• L-glutamine• HEPES
• Phenol Red• Sodium Bicarbonate

The complete formulation is available.

Using RPMI 1640
RPMI 1640 Medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 Medium contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. RPMI 1640 Medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 Medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 Medium uses a sodium bicarbonate buffer system (2.0 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH. Powder forms of Gibco™ cell culture media require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).

Product use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. Customers using Gibco™ RPMI 1640 in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

cGMP manufacturing and quality system
RPMI 1640 Medium is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer an identical RPMI 1640 product made in our Scotland facility (51800-043). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

This product is not used for in vitro diagnostic purpose in some countries.
Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. Customers using Gibco RPMI 1640 in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from Life Technologies to reference their Type II Drug Master File (DMF). For in vitro diagnostic use.
사양
세포주HeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas
세포 유형Leukemic Cells
제조 품질cGMP-compliant under the ISO 13485 standard
제품라인Gibco
제품 유형RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium)
수량10 L
유통 기한36 Months From Date of Manufacture
배송 조건Room Temperature
분류Animal Origin-free
형태Powder
Serum LevelStandard Serum Supplementation
멸균Sterile-filtered
첨가제 포함Glutamine
첨가제 없음No Sodium Pyruvate
Unit SizeEach
구성 및 보관
Storage conditions: 2-8° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture

자주 묻는 질문(FAQ)

How light sensitive is RPMI 1640 media? Should I also be protecting it from LED light?

While we know that different wavelengths of light are worse than others for exposure, we would recommend as a best practice to protect the medium from all forms of light exposure including LEDs, as much as possible to ensure optimal performance, as several components within the medium are light sensitive, such as vitamins.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the density (g/L) for RPMI 1640 Medium?

We have specific gravity information for RPMI 1640 Medium: 1.006 kg/L. In this case, the specific gravity is the same as density as the solvent is water.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the shelf life of my powdered media once I reconstitute it?

Expiration date of most Gibco reconstituted dry format media (AGT or DPM) has not been established; end users should assess performance and stability of this reconstituted media in a system that is relevant to their process. A dry format product (either DPM or AGT) will age upon storage and while we may not be able to detect which component(s) degrade since we can't measure every component, that doesn't mean that the reconstituted liquid will have the same stability as a fresh liquid made by direct weigh methods.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.