Indium, 99.99%, (trace metal basis), foil, 0.127 mm thick
Indium, 99.99%, (trace metal basis), foil, 0.127 mm thick
Indium, 99.99%, (trace metal basis), foil, 0.127 mm thick
Thermo Scientific Chemicals

Indium, 99.99%, (trace metal basis), foil, 0.127 mm thick

CAS: 7440-74-6 | In | 114.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.3 g
9.2 g
Catalog number 318110023
Price (EUR)
136,00
Each
Quantity:
2.3 g
Request bulk or custom format
Price (EUR)
136,00
Each
Chemical Identifiers
CAS7440-74-6
IUPAC Nameindium
Molecular FormulaIn
InChI KeyAPFVFJFRJDLVQX-UHFFFAOYSA-N
SMILES[In]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Foil
Total trace metal impurities=<0.01 %
Appearance (Color)Grey
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only