Antimony tribromide, 97%, pure
Antimony tribromide, 97%, pure
Antimony tribromide, 97%, pure
Thermo Scientific Chemicals

Antimony tribromide, 97%, pure

CAS: 7789-61-9 | Br3Sb | 361.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 318400500
Price (USD)
111.65
Special offer
Online exclusive
Ends: 30-Jun-2026
131.00
Save 19.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
111.65
Special offer
Online exclusive
Ends: 30-Jun-2026
131.00
Save 19.35 (15%)
Each
Chemical Identifiers
CAS7789-61-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Titration Argentometric>=96.0 %
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only